Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001979.2 | NP_001001979.1 | 20 | 3250 | ||
GenBank mRNA | AK147238.1 | - | 20 | 3250 | ||
AK148084.1 | - | 20 | 2813 | |||
AK166316.1 | - | 20 | 2847 | |||
BC075647.1 | - | 20 | 2865 | 30620548 |