Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027436.3 | NP_081712.2 | 13 | 1617 | ||
GenBank mRNA | AK017491.1 | - | 13 | 1593 | ||
AK038479.1 | - | 13 | 1611 | |||
AK044460.1 | - | 13 | 1602 | |||
AK076613.1 | - | 13 | 1547 | |||
AK146428.1 | - | 14 | 1608 | |||
AK183152.1 | - | 2 | 195 | |||
AK218084.1 | - | 2 | 249 | |||
AK219345.1 | - | 1 | 157 | |||
BC049871.1 | - | 4 | 457 | |||
BC110358.1 | - | 13 | 1595 | 5322591 | ||
BC144798.1 | - | 13 | 1598 | |||
BC145711.1 | - | 13 | 1598 | 40131014 |