Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027436.3 | NP_081712.2 | 7 | 946 | ||
GenBank mRNA | AK017491.1 | - | 7 | 922 | ||
AK038479.1 | - | 7 | 940 | |||
AK044460.1 | - | 7 | 931 | |||
AK076613.1 | - | 7 | 874 | |||
AK146428.1 | - | 8 | 937 | |||
BC110358.1 | - | 7 | 924 | 5322591 | ||
BC144798.1 | - | 7 | 927 | |||
BC145711.1 | - | 7 | 927 | 40131014 |