Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001321.3 | NP_001001321.2 | 12 | 915 | ||
GenBank mRNA | AB117931.1 | - | 12 | 896 | ||
AB117932.1 | - | 11 | 813 | |||
AK154257.1 | - | 12 | 941 | |||
AK159148.1 | - | 12 | 916 | |||
BC046402.1 | - | 6 | 455 | |||
BC107210.1 | - | 11 | 862 | |||
BC107211.1 | - | 12 | 895 | 40051417 |