Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027452.3 | NP_081728.2 | 2 | 1602 | ||
GenBank mRNA | AK017594.1 | - | 2 | 1806 | ||
AK044375.1 | - | 2 | 1479 | |||
AK122476.1 | - | 2 | 1557 | |||
BC139418.1 | - | 2 | 1602 | 8862440 |