Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178395.3 | NP_848482.1 | 7, 8 | 829 | ||
GenBank mRNA | AK018284.1 | - | 801 | |||
AK030662.1 | - | 7, 8 | 830 | |||
AK044652.1 | - | 7, 8 | 823 | |||
AK046533.1 | - | 7, 8 | 829 | |||
AK136296.1 | - | 7, 8 | 828 | |||
BC117761.1 | - | 8, 9 | 591 | |||
BC127156.1 | - | 8, 9 | 663 | 40090859 |