Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081368.1 | NP_001074837.1 | 2 | 540 | ||
GenBank mRNA | AK017705.1 | - | 2 | 547 | ||
AK035909.1 | - | 2 | 1106 | |||
AK039905.1 | - | 2 | 365 | |||
AK040787.1 | - | 2 | 512 | |||
AK043059.1 | - | 2 | 514 | |||
AK133804.1 | - | 2 | 480 | |||
AK179171.1 | - | 1 | 75 | |||
AK196414.1 | - | 87 | ||||
BC082558.1 | - | 2 | 509 | 30605020 | ||
CF749564.1 | - | 2 | 640 |