Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029617.2 | NP_083893.2 | 16 | 5690 | ||
GenBank mRNA | AK017750.1 | - | 4 | 339 |