Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Medag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027519.3 | NP_081795.1 | 5 | 1034 | ||
GenBank mRNA | AB055406.1 | - | 5 | 1065 | ||
AK018128.1 | - | 5 | 850 | |||
AK036605.1 | - | 5 | 1035 | |||
AK042557.1 | - | 5 | 747 | |||
AK047657.1 | - | 5 | 1091 | |||
AK086092.1 | - | 5 | 1043 | |||
AK138268.1 | - | 5 | 733 | |||
AK160904.1 | - | 5 | 1041 | |||
AK161526.1 | - | 5 | 1043 | |||
AK162725.1 | - | 5 | 1035 | |||
AK163617.1 | - | 5 | 733 | |||
AK165529.1 | - | 5 | 831 | |||
BC026658.1 | - | 5 | 797 | |||
BC116246.1 | - | 5 | 956 | 40093530 | ||
BC116247.1 | - | 5 | 954 | 40093536 |