Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027541.4 | NP_081817.2 | 10, 11 | 1912 | ||
GenBank mRNA | AK014398.1 | - | 10, 11 | 1520 | ||
AK088194.1 | - | 10, 11 | 1913 | |||
AK139852.1 | - | 10, 11 | 1545 | |||
AK167435.1 | - | 10, 11 | 1549 | |||
BC006782.1 | - | 10, 11 | 1497 | 3589226 | ||
BC026607.1 | - | 7, 8 | 1666 | |||
BC040787.1 | - | 7, 8 | 1666 |