Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033158.2 | NP_001028330.2 | 4 | 485 | ||
NM_001162923.1 | NP_001156395.1 | 4 | 485 | |||
GenBank mRNA | AK014511.1 | - | 4 | 448 | ||
AK048176.1 | - | 4 | 439 | |||
AK087136.1 | - | 5 | 973 | |||
AK171623.1 | - | 4 | 441 | |||
BC125036.1 | - | 4 | 462 | 40102054 |