Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025428.1 | NP_001020599.1 | 25 | 3324 | ||
NM_001025429.1 | NP_001020600.1 | 25 | 3321 | |||
NM_001025430.1 | NP_001020601.1 | 24 | 3309 | |||
NM_001199203.1 | NP_001186132.1 | 28 | 3800 | |||
NM_001199204.1 | NP_001186133.1 | 28 | 3797 | |||
NM_001199205.1 | NP_001186134.1 | 25 | 3348 | |||
NM_027569.2 | NP_081845.2 | 27 | 3758 | |||
GenBank mRNA | AB047782.1 | - | 28 | 3720 | ||
AF327451.1 | - | 27 | 3740 | |||
AK049161.1 | - | 19 | 2360 | |||
AK147431.1 | - | 25 | 3405 | |||
AK147537.1 | - | 25 | 3424 | |||
AK172961.1 | - | 25 | 3303 | |||
AY823270.1 | - | 27 | 3302 | |||
BC052058.1 | - | 25 | 3259 | |||
BC060100.1 | - | 24 | 3243 | 6417257 | ||
BC060506.1 | - | 13 | 1681 | |||
BC094670.1 | - | 24 | 3237 | 30618738 | ||
FJ042496.1 | - | 28 | 3674 |