Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023596.3 | NP_076085.1 | 4 | 484 | ||
GenBank mRNA | AF326986.1 | - | 4 | 495 | ||
AK017065.1 | - | 1 | 380 | |||
AK034133.1 | - | 4 | 480 | |||
AK051317.1 | - | 4 | 509 | |||
AK157868.1 | - | 4 | 485 | |||
BC137864.1 | - | 4 | 467 | 8860883 | ||
BC144983.1 | - | 4 | 467 | |||
CV557129.1 | - | 3 | 354 |