Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mamdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174857.3 | NP_777282.1 | 9 | 1448 | ||
GenBank mRNA | AK004794.1 | - | 8 | 1311 | ||
AK035566.1 | - | 9 | 1449 | |||
AK154219.1 | - | 9 | 1525 | |||
BC042773.1 | - | 9 | 1477 | 4019075 |