Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163617.1 | NP_001157089.1 | 2 | 341 | ||
NM_001163618.1 | NP_001157090.1 | 2 | 341 | |||
NM_001163619.1 | NP_001157091.1 | 2 | 341 | |||
NM_027931.3 | NP_082207.2 | 2 | 341 | |||
GenBank mRNA | AK011540.1 | - | 2 | 227 | ||
AK080690.1 | - | 2 | 244 | |||
AK088675.1 | - | 2 | 216 | |||
AK142685.1 | - | 2 | 236 | |||
AK164589.1 | - | 2 | 248 | |||
AK164825.1 | - | 2 | 199 | |||
AK168075.1 | - | 2 | 235 | |||
AK190261.1 | - | 1 | 13 | |||
BC008995.1 | - | 2 | 226 | 3157108 | ||
BC147501.1 | - | 2 | 225 | 9056383 | ||
BG087111.2 | - | 2 | 193 |