Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027951.1 | NP_082227.1 | 6 | 1391 | ||
GenBank mRNA | AK005842.1 | - | 6 | 1393 | ||
AY485267.1 | - | 6 | 1391 | |||
BC117510.1 | - | 6 | 1344 | 40105676 | ||
BC117526.1 | - | 6 | 1344 | 40105675 |