Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Aox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023631.2 | NP_076120.2 | 34 | 4098 | ||
GenBank mRNA | AF233581.1 | - | 34 | 4025 | ||
AK009112.1 | - | 9 | 1017 | |||
AK158427.1 | - | 34 | 4099 | |||
BC117975.1 | - | 34 | 4025 | 40098789 | ||
BC117976.1 | - | 34 | 4025 | 40098791 |