Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027982.2 | NP_082258.2 | 3 | 671 | ||
GenBank mRNA | AK009235.1 | - | 3 | 610 | ||
AY184802.1 | - | 3 | 671 | |||
BC117498.1 | - | 3 | 667 | 40106734 | ||
BC121788.1 | - | 3 | 666 | 40106732 |