Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028047.2 | NP_082323.1 | 10, 11 | 1374 | ||
GenBank mRNA | AK005126.1 | - | 10, 11 | 1374 | ||
AK039368.1 | - | 10, 11 | 1373 | |||
AK141576.1 | - | 10, 11 | 1365 | |||
BC034211.1 | - | 10, 11 | 1289 | 4988361 |