Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028047.2 | NP_082323.1 | 9 | 1230 | ||
GenBank mRNA | AK005126.1 | - | 9 | 1230 | ||
AK039368.1 | - | 9 | 1229 | |||
AK141576.1 | - | 9 | 1221 | |||
AK169651.1 | - | 9 | 1154 | |||
BC034211.1 | - | 9 | 1145 | 4988361 |