Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mccc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023644.4 | NP_076133.3 | 17 | 2018 | ||
GenBank mRNA | AF310338.1 | - | 17 | 1939 | ||
AK007782.1 | - | 17 | 1971 | |||
AK031072.1 | - | 17 | 1979 | |||
AK032203.1 | - | 17 | 1977 | |||
AK141040.1 | - | 2 | 4826 | |||
AK168589.1 | - | 17 | 1907 | |||
BC021382.1 | - | 17 | 1982 | 5042993 |