Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mccc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023644.4 | NP_076133.3 | 11 | 1314 | ||
GenBank mRNA | AF310338.1 | - | 11 | 1235 | ||
AK007782.1 | - | 11 | 1267 | |||
AK031072.1 | - | 11 | 1275 | |||
AK032203.1 | - | 11 | 1257 | |||
AK160184.1 | - | 9 | 1031 | |||
AK168589.1 | - | 11 | 1203 | |||
AK199182.1 | - | 2 | 261 | |||
BC021382.1 | - | 11 | 1278 | 5042993 |