Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285839.1 | NP_001272768.1 | 6 | 1376 | ||
GenBank mRNA | AK011265.1 | - | 6 | 1324 | ||
AK045669.1 | - | 6 | 1338 | |||
AK137332.1 | - | 7 | 1407 | |||
BC038910.1 | - | 4 | 702 | |||
BC051211.1 | - | 3 | 752 | |||
BC058172.1 | - | 6 | 1339 | 5012054 |