Gene Symbol: Tmem68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028097.3 | NP_082373.1 | 3 | 369 | ||
GenBank mRNA | AK008493.1 | - | 3 | 403 | ||
AK048210.1 | - | 3 | 369 | |||
AK084881.1 | - | 3 | 403 | |||
AK088600.1 | - | 3 | 412 | |||
AK147765.1 | - | 3 | 402 | |||
AK162563.1 | - | 3 | 402 | |||
AK165916.1 | - | 3 | 431 | |||
AK166237.1 | - | 3 | 455 | |||
BC016240.1 | - | 3 | 387 | 4506688 |