Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028130.3 | NP_082406.2 | 5 | 1319 | ||
GenBank mRNA | AB010336.1 | - | 1 | 129 | ||
AK028290.1 | - | 5 | 1280 | |||
BC059261.1 | - | 5 | 1291 | 6409233 |