Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028136.2 | NP_082412.2 | 8 | 1133 | ||
GenBank mRNA | AF448804.1 | - | 8 | 1133 | ||
AK013031.1 | - | 1 | 125 | |||
AK129373.1 | - | 8 | 1112 | |||
BC004634.1 | - | 6 | 724 | |||
BC006615.1 | - | 8 | 1133 | |||
BC064890.1 | - | 8 | 1110 | |||
BC138061.1 | - | 8 | 1109 | 8861080 |