Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033165.3 | NP_001028337.2 | 3 | 972 | ||
GenBank mRNA | AK136359.1 | - | 3 | 666 | ||
BC119039.1 | - | 3 | 737 | 8733788 | ||
BC120693.1 | - | 3 | 737 | 40129616 | ||
BX512063.1 | - | 1 | 262 |