Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025392.1 | NP_001020563.1 | 6, 7 | 2086 | ||
NM_001025393.1 | NP_001020564.1 | 6, 7 | 2086 | |||
NM_153787.2 | NP_722482.1 | 6, 7 | 2080 | |||
GenBank mRNA | AK088741.1 | - | 6, 7 | 2021 | ||
AK129071.1 | - | 3, 4 | 1370 | |||
AK141834.1 | - | 6, 7 | 2033 | |||
AK149724.1 | - | 6, 7 | 2039 | |||
AK150017.1 | - | 6, 7 | 2039 | |||
AK161608.1 | - | 975 | ||||
AK161692.1 | - | 6, 7 | 2084 | |||
BC029682.1 | - | 6, 7 | 2017 | |||
BC034300.1 | - | 6, 7 | 2012 | 4507876 | ||
BC086624.1 | - | 6, 7 | 2068 |