Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adipor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012847.1 | - | 7 | 1062 | ||
AK143679.1 | - | 7 | 1455 | |||
AK143680.1 | - | 1150 | ||||
AK148140.1 | - | 7 | 1024 | |||
AK151565.1 | - | 7 | 1037 | |||
AK167896.1 | - | 7 | 1015 | |||
AK168069.1 | - | 7 | 1051 | |||
AK180314.1 | - | 3 | 296 | |||
AK185902.1 | - | 2 | 58 | |||
AK187788.1 | - | 1 | 37 | |||
AK189666.1 | - | 2 | 37 | |||
AK191648.1 | - | 2 | 37 | |||
AK200337.1 | - | 2 | 37 | |||
AY424290.1 | - | 7 | 1026 | |||
BC014875.1 | - | 7 | 1026 | 2811173 |