Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adipor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137258.1 | - | 3, 4 | 1212 | ||
AK143679.1 | - | 4, 5 | 1049 | |||
AK143680.1 | - | 744 | ||||
AK148140.1 | - | 4, 5 | 618 | |||
AK151565.1 | - | 4, 5 | 631 | |||
AK167896.1 | - | 4, 5 | 609 | |||
AK168069.1 | - | 4, 5 | 645 | |||
AK214372.1 | - | 1, 2 | 150 | |||
AY424290.1 | - | 4, 5 | 620 | |||
BC014875.1 | - | 4, 5 | 620 | 2811173 |