Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164583.1 | NP_001158055.1 | 16 | 2338 | ||
NM_001164584.1 | NP_001158056.1 | 16 | 2283 | |||
NM_001164585.1 | NP_001158057.1 | 17 | 2348 | |||
NM_198412.2 | NP_940804.1 | 17 | 2257 | |||
GenBank mRNA | AK012831.1 | - | 7 | 1003 | ||
AK049935.1 | - | 4 | 300 | |||
AK122293.1 | - | 16 | 2259 | |||
AK147570.1 | - | 17 | 2220 | |||
AK147637.1 | - | 17 | 2220 | |||
AK163657.1 | - | 16 | 2157 | |||
BC035262.1 | - | 11 | 1492 | |||
BC060734.1 | - | 16 | 2338 | 6409942 | ||
BC064460.1 | - | 17 | 2226 | 6821822 |