Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef10l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112722.1 | NP_001106193.1 | 28 | 3586 | ||
NM_001112723.1 | NP_001106194.1 | 27 | 3544 | |||
NM_001290803.1 | NP_001277732.1 | 29 | 3685 | |||
NM_172415.3 | NP_766003.3 | 28 | 3661 | |||
GenBank mRNA | AK028570.1 | - | 22 | 3002 | ||
AK028648.1 | - | 28 | 3569 | |||
AK129408.1 | - | 27 | 3544 | |||
BC006747.1 | - | 1 | 225 | |||
BC021843.1 | - | 14 | 1831 | |||
BC024646.1 | - | 3 | 576 | |||
BC034061.1 | - | 22 | 2822 | |||
BC059220.1 | - | 28 | 3649 | 6417641 |