Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176860.5 | NP_789830.1 | 10 | 1461 | ||
GenBank mRNA | AB075602.1 | - | 10 | 1456 | ||
AK013361.1 | - | 10 | 1529 | |||
AK034450.1 | - | 10 | 1652 | |||
AK035764.1 | - | 10 | 1462 | |||
AK133948.1 | - | 10 | 1459 | |||
AK150909.1 | - | 10 | 2028 | |||
AK151721.1 | - | 10 | 2029 | |||
AK151895.1 | - | 10 | 1662 | |||
AK152013.1 | - | 10 | 1662 | |||
AK154576.1 | - | 10 | 1683 | |||
BC053436.1 | - | 10 | 1433 | 6510702 |