Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spock3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252620.1 | NP_001239549.1 | 9 | 957 | ||
NM_001252621.1 | NP_001239550.1 | 8 | 948 | |||
NM_023689.3 | NP_076178.1 | 9 | 1071 | |||
GenBank mRNA | AJ278998.1 | - | 9 | 965 | ||
AK013644.1 | - | 9 | 1000 | |||
AK049843.1 | - | 8 | 949 | |||
AK140736.1 | - | 2 | 258 | |||
AK220280.1 | - | 9 | 1055 | |||
BC017601.1 | - | 9 | 970 | 4502169 | ||
BC053334.1 | - | 9 | 942 | 6492113 |