Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm7sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028454.2 | NP_082730.2 | 4 | 578 | ||
GenBank mRNA | AF480070.1 | - | 4 | 452 | ||
AK014166.1 | - | 4 | 559 | |||
AK037325.1 | - | 3 | 382 | |||
AK039028.1 | - | 4 | 508 | |||
AK040016.1 | - | 4 | 454 | |||
AK043687.1 | - | 3 | 761 | |||
AK134512.1 | - | 1 | 1279 | |||
AK155475.1 | - | 4 | 535 | |||
BC014769.1 | - | 4 | 570 | 4239159 |