Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028568.1 | NP_082844.1 | 1 | 300 | ||
GenBank mRNA | AK007061.1 | - | 1 | 313 | ||
BC048605.1 | - | 1 | 300 | 6741358 |