Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028599.2 | NP_082875.1 | 16 | 1759 | ||
GenBank mRNA | AK010772.1 | - | 16 | 1759 | ||
AK152412.1 | - | 16 | 1760 | |||
AK163221.1 | - | 16 | 1733 | |||
AK168032.1 | - | 16 | 1753 | |||
AK168163.1 | - | 16 | 1761 | |||
BC038920.1 | - | 2 | 76 | |||
BC049925.1 | - | 4 | 1217 | |||
BC055483.1 | - | 5 | 660 | |||
BC085177.1 | - | 16 | 1789 | |||
BC103800.1 | - | 2 | 92 | |||
BC117880.2 | - | 16 | 1789 | 40096010 | ||
BC117881.1 | - | 16 | 1786 | 40096014 |