Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028748.2 | NP_083024.1 | 7 | 835 | ||
GenBank mRNA | AK002481.1 | - | 7 | 805 | ||
AK035475.1 | - | 7 | 847 | |||
AK052776.1 | - | 4 | 386 | |||
AK085411.1 | - | 7 | 836 | |||
AK086679.1 | - | 7 | 849 | |||
AY424294.1 | - | 7 | 805 | |||
BC054855.1 | - | 7 | 861 | |||
BC100516.1 | - | 6 | 716 | 30919009 |
Set Membership: |
![]() |