Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028762.1 | NP_083038.1 | 16, 17 | 2151 | ||
GenBank mRNA | AK004657.1 | - | 7, 8 | 966 | ||
AK146493.1 | - | 16, 17 | 2184 | |||
AK167100.1 | - | 16, 17 | 2148 | |||
BC025619.1 | - | 16, 17 | 2151 | 5320947 | ||
BC034010.1 | - | 16, 17 | 2117 | 4225239 |