Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Robo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF536772.1 | - | 2 | 191 | ||
AK004723.2 | - | 2 | 188 | |||
AK087355.1 | - | 2 | 185 | |||
AK172104.1 | - | 2 | 338 |