Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028816.2 | NP_083092.2 | 12 | 2086 | ||
GenBank mRNA | AB093235.1 | - | 7 | 2351 | ||
AK049009.1 | - | 12 | 2099 | |||
AK154709.1 | - | 12 | 2131 | |||
AY029528.1 | - | 11 | 1582 | |||
BC058090.1 | - | 12 | 2086 | 6826356 |