Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286007.1 | NP_001272936.1 | 12 | 2217 | ||
NM_001286009.1 | NP_001272938.1 | 12 | 2217 | |||
NM_001286011.1 | NP_001272940.1 | 11 | 1984 | |||
NM_133755.2 | NP_598516.2 | 12 | 2144 | |||
GenBank mRNA | AK006233.1 | - | 2 | 248 | ||
AK033157.1 | - | 12 | 1934 | |||
AK045084.1 | - | 12 | 1933 | |||
AK146771.1 | - | 12 | 2139 | |||
AK153511.1 | - | 12 | 2065 | |||
AK158060.1 | - | 12 | 1933 | |||
AK167924.1 | - | 12 | 2039 | |||
BC012519.1 | - | 12 | 2072 | 3601589 | ||
BC025564.1 | - | 11 | 1906 | |||
BC025582.1 | - | 12 | 2033 | 5324981 | ||
BC027034.1 | - | 11 | 2418 |