Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177589.3 | NP_808257.2 | 21 | 2316 | ||
GenBank mRNA | AK016524.1 | - | 21 | 2315 | ||
AK043628.1 | - | 22 | 2371 | |||
AK132731.1 | - | 21 | 2317 | |||
BC065094.1 | - | 22 | 2614 | |||
BC109364.2 | - | 12 | 1286 | 40059232 | ||
BC109365.2 | - | 12 | 1251 | 40059233 |