Search Thermo Fisher Scientific
Gene Symbol: Taf7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028958.4 | NP_083234.2 | 10, 11 | 1222 | ||
GenBank mRNA | AF285574.1 | - | 10, 11 | 1026 | ||
AK017109.1 | - | 10, 11 | 1223 | |||
BC106854.2 | - | 10, 11 | 1014 | 40046501 |