Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam46c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142952.1 | NP_001136424.1 | 2 | 251 | ||
GenBank mRNA | AK015259.1 | - | 1 | 55 | ||
BC037072.1 | - | 2 | 241 | |||
BC046309.1 | - | 2 | 245 | |||
BC120717.1 | - | 2 | 242 | 40129640 | ||
BC120719.1 | - | 2 | 242 | 40129642 |