Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029044.2 | NP_083320.1 | 7, 8 | 900 | ||
GenBank mRNA | AK015430.1 | - | 7, 8 | 902 | ||
AK049695.1 | - | 7, 8 | 928 | |||
BC018412.1 | - | 7, 8 | 913 | 3673669 |