Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147218.2 | NP_671751.2 | 25, 26 | 3615 | ||
GenBank mRNA | AF498361.1 | - | 25, 26 | 3567 | ||
BC132417.1 | - | 25, 26 | 3452 | 40130694 | ||
BC132419.1 | - | 25, 26 | 3452 | 40130696 |