Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erp44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029572.2 | NP_083848.1 | 3, 4 | 325 | ||
GenBank mRNA | AK003217.1 | - | 3, 4 | 294 | ||
AK151497.1 | - | 3, 4 | 326 | |||
AK160113.1 | - | 3, 4 | 308 | |||
AK172973.1 | - | 267 | ||||
BC019558.1 | - | 3, 4 | 232 | 4192664 |