Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Erp44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029572.2 | NP_083848.1 | 8 | 822 | ||
GenBank mRNA | AK003217.1 | - | 8 | 791 | ||
AK141377.1 | - | 8 | 780 | |||
AK151497.1 | - | 8 | 823 | |||
AK160113.1 | - | 8 | 805 | |||
AK172973.1 | - | 764 | ||||
BC019558.1 | - | 8 | 729 | 4192664 |