Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029576.3 | NP_083852.1 | 1, 2 | 76 | ||
GenBank mRNA | AB232584.1 | - | 18 | |||
AK003609.1 | - | 1, 2 | 77 | |||
AK042020.1 | - | 1, 2 | 78 | |||
AK156726.1 | - | 1, 2 | 99 | |||
BC016408.1 | - | 1, 2 | 71 | 4242206 |